Learn More
Sino Biological SARS-CoV-2 (BA.2.12.1) Spike RBD Protein (His Tag) 100ug
Supplier:  Sino Biological 40592V08H132100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (BA.2.12.1) Spike RBD (YP_009724390.1, with mutations G339D, S371F, S373P, S375F, T376A, D405N, R408S, K417N, N440K, L452Q, S477N, T478K, E484A, Q493R, Q498R, N501Y, Y505H)(Arg319-Phe541) was expressed with a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant BA.2.12.1).
Catalog No. 50-224-6499