Learn More
Sino Biological SARS-CoV-2 (C.37) Spike S1+S2 trimer Protein ( ECD, His Tag) 100ug
Supplier:  Sino Biological 40589V08H24100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike S1+S2 (YP_009724390.1, with mutations G75V, T76I, R246N, SYLTPGD247-253del, L452Q, F490S, D614G, T859N,F817P, A892P, A899P,A942P, K986P, V987P and furin cleavage site mutants) expressed with the bacteriophage T4 fibritin and a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant C.37) which emerged in the Peru.
Catalog No. 50-224-6478