Learn More
Sino Biological SARS-CoV-2 Delta Spike RBD Specific Antibody, Chimeric MAb
Supplier:  Sino Biological 40589D006100
Encompass_Preferred
It is a chimeric monoclonal antibody combining the constant domains of the human IgG1 molecule with mouse variable regions. The variable region was obtained from a mouse immunized with purified, recombinant SARS-CoV-2 Delta Spike ECD Protein. The antibody was produced using recombinant antibody technology.
Catalog No. 50-228-2700