Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ MAS™ SARS-CoV-2 Serology Quality Controls
Click to view available options
Type:
SARS-CoV-2 IgG Positive Control
SARS-CoV-2 Negative Control
Description
Assess the performance of serological assays for SARS-CoV-2 infections using the Thermo Scientific MAS SARS-CoV-2 Serology Quality Controls. These third-party controls include individual positive control for SARS-CoV-2 as well as a negative control for accuracy.
- 21 days open vial stability at 2°C to 8°C
- Ready to use straight from the refrigerator
- Human Plasma matrix
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Detectable Analytes | SARS-CoV-2 Virus IgG Ab |
| Type | SARS-CoV-2 IgG Positive Control |
| Packaging | One box of control contains five tubes SARS-CoV-2 IgG Positive Control |
| Quantity | 5 x 3.0 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction