Learn More
Cayman Chemical SARSCoV2 InhIbItr Control 1ea
SDP
Supplier:  Cayman Chemical 4020551 EA
Encompass_Preferred
SARS-CoV-2 Inhibitor Control has been tested and formulated to work exclusively with Cayman’s SARS-CoV-2 Spike-ACE2 Interaction Inhibitor Screening Assay Kit
Catalog No. 50-294-2156