Learn More
Cayman Chemical SARS-CoV-2 Main Protease Assay Buffer, 1 ea
SDP
Catalog No. 502893158
Encompass_Preferred
SARS-CoV-2 Main Protease Assay Buffer has been tested and formulated to work exclusively with Cayman’s SARS-CoV-2 Main Protease Inhibitor Screening Assay Kit (Item No. 701960).
Catalog No. 50-289-3158 Supplier Cayman Chemical Supplier No. 7019611 EA
SARS-CoV-2 Main Protease Assay Buffer has been tested and formulated to work exclusively with Cayman’s SARS-CoV-2 Main Protease Inhibitor Screening Assay Kit (Item No. 701960).