Learn More
Epigentek Inc SARS-CoV-2 N Recombinant Antibody (100ul)
SDP
Supplier:  Epigentek Inc A73676100
Encompass_Preferred
HRP Conjugated Recombinant Antibody Raised in Mouse. Immunogen: Recombinant Human Novel Coronavirus Nucleoprotein (N) (1-419aa). Tested Applications: ELISA. Reactivity: SARS-CoV-2. Alternative Names: Nucleocapsid protein, NC, protein N, N.
Catalog No. 50-201-9335