Learn More
Cayman Chemical SARSCoV2 nsp10recombInt 500ug
SDP
Catalog No. 502943184
Encompass_Preferred
Recombinant SARS-CoV-2 N-terminal His-tagged nsp10 expressed in E. coli • Amino acids: 1-139 (full length)
Catalog No. 50-294-3184 Supplier Cayman Chemical Supplier No. 40883500 UG
Recombinant SARS-CoV-2 N-terminal His-tagged nsp10 expressed in E. coli • Amino acids: 1-139 (full length)