Learn More
Cayman Chemical SARSCoV2 nsp13 HelIcaseE 10ug
SDP
Catalog No. 502991317
Encompass_Preferred
Source: Active recombinant nsp13 helicase expressed in E. coli • Amino acids: 5325-5925 • MW: 69.1 kDa
Catalog No. 50-299-1317 Supplier Cayman Chemical Supplier No. 3058910 UG
Source: Active recombinant nsp13 helicase expressed in E. coli • Amino acids: 5325-5925 • MW: 69.1 kDa