Learn More
Cayman Chemical SARSCoV2 nsp14 Methyltrnsf
SDP
Catalog No. 502942533
Encompass_Preferred
Source: Recombinant SARS-CoV-2 N-terminal GST-tagged nsp14 expressed in E. coli • Amino acids: 1-527 (full length) • MW: 86.3 kDa
Catalog No. 50-294-2533 Supplier Cayman Chemical Supplier No. 40884500 UG
Source: Recombinant SARS-CoV-2 N-terminal GST-tagged nsp14 expressed in E. coli • Amino acids: 1-527 (full length) • MW: 86.3 kDa