Learn More
Cayman Chemical SARSCoV2 nsp16 Methyltrnsf
SDP
Supplier:  Cayman Chemical 31816100 UG
Encompass_Preferred
Source: Recombinant C-terminal His-tagged SARS-CoV-2 nsp16 methyltransferase expressed in E. coli • Amino acids: 6799-7096 • MW: 33.5 kDa
Catalog No. 50-294-7305