Learn More
Epigentek Inc SARS-CoV-2 Nucleocapsid Protein, Avi-His-tag (1 mg)
SDP
Supplier:  Epigentek Inc E800271
Encompass_Preferred
SARS-CoV-2 nucleocapsid protein, also known as COVID-19 nucleocapsid and SARS-CoV-2 N protein, Genbank Accession No.: YP_009724397.2, a.a. 1-419(end), with C-terminal Avi-His-tag, expressed in a HEK293 cell expression system. MW= 48 kDa. This protein runs at a higher M.W. by SDS-PAGE due to glycosylation.
Catalog No. 50-201-9388