Learn More
American Research Products Inc SARSCoV2 Nucleocapsid
Supplier:  American Research Products Inc IVMB0241500UG
Encompass
VivoGenie SARS-CoV-2 Nucleocapsid (Anti-N) Low Endotoxin Antibody is a vivogenie with sars-cov-2 virus reactivity Available in 100 ug size with 7 - 14 business days lead time Ships via gel pack Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month For longer term storage aseptically aliquot in working volumes without diluting and store at -80 C Avoid Repeated Freeze Thaw Cycles
Catalog No. 50-003-05179