Learn More
Sino Biological SARS-CoV-2 Nucleocapsid Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 68097R150100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant SARS-CoV-2 Nucleocapsid CTD Protein (Catalog#40588-V07E5; YP_009724397.2; Lys248-Pro365).
Catalog No. 50-224-6546