Learn More
Sino Biological SARS-CoV-2 Nucleocapsid Antibody, Mouse MAb
Supplier:  Sino Biological 40588MM162100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant SARS-CoV-2 Nucleocapsid Protein (Catalog#40588-V08B; YP_009724397.2; Met1-Ala419(335Gly/Ala)). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-228-2413