Learn More
Sino Biological SARS-CoV-2 Nucleocapsid (D63G, R203M, D377Y) Protein (His Tag) 100ug
Supplier:  Sino Biological 40588V07E29100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Nucleocapsid (YP_009724397.2, with mutations D63G, R203M, D377Y) (Met1-Ala419) was expressed with a polyhistidine tag at the N-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant B.1.617.2) which emerged in the India.
Catalog No. 50-211-4302
 Restricted Item