Learn More
Sino Biological SARS-CoV-2 Nucleocapsid (P13L, R203K, G204R, Q384H) Protein (His Tag)
Supplier:  Sino Biological 40588V07E33100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Nucleocapsid (YP_009724397.2, with mutations P13L, R203K, G204R, Q384H) (Met1-Ala419) was expressed with a polyhistidine tag at the N-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant C.1.2) which emerged in the South Africa.
Catalog No. 50-224-7159