Learn More
Cayman Chemical SARSCoV2 NuclocapsId Prot 50ug
SDP
Catalog No. 502992131
Encompass_Preferred
Amino acids: 1-419 (full length) • MW: 49.7 kDa
Catalog No. 50-299-2131 Supplier Cayman Chemical Supplier No. 3042750 UG
Amino acids: 1-419 (full length) • MW: 49.7 kDa