Learn More
Cayman Chemical SARSCoV2 nutralIzng ntIb 500uL
SDP
Catalog No. 502894105
Encompass_Preferred
An NSAID and COX inhibitor (IC50s 7 and 4 UM for COX-1 and COX-2 respectively) reduces paw volume in a rat model of adjuvant-induced polyarthritis at 3 15 and 75 mg/kg induces antinociception in rats (ED50 54.3 mg/kg) reduces carrageenan-induced paw edema in rats at 20-180 mg/kg
Catalog No. 50-289-4105 Supplier Cayman Chemical Supplier No. 502073500 UL
An NSAID and COX inhibitor (IC50s 7 and 4 UM for COX-1 and COX-2 respectively) reduces paw volume in a rat model of adjuvant-induced polyarthritis at 3 15 and 75 mg/kg induces antinociception in rats (ED50 54.3 mg/kg) reduces carrageenan-induced paw edema in rats at 20-180 mg/kg