Learn More
Cayman Chemical SARSCoV2 nutralIzng ntIb 50uL
SDP
Catalog No. 502990710
Encompass_Preferred
An N-acyl amide increases the OCR of mitochondria isolated from mouse brown adipose tissue in a Seahorse assay and decreases oligomycin-induced increases in the mitochondrial membrane potential in C2C12 cells indicating mitochondrial uncoupling activity at 50 UM reduces food intake and body weight of diet-induced obese mice at 30 mg/kg for 8 days
Catalog No. 50-299-0710 Supplier Cayman Chemical Supplier No. 3156950 UL
An N-acyl amide increases the OCR of mitochondria isolated from mouse brown adipose tissue in a Seahorse assay and decreases oligomycin-induced increases in the mitochondrial membrane potential in C2C12 cells indicating mitochondrial uncoupling activity at 50 UM reduces food intake and body weight of diet-induced obese mice at 30 mg/kg for 8 days