Learn More
Cayman Chemical SARS-CoV-2 Neutralizing Antibody Detection ELISA Kit, 96 Well
SDP
Catalog No. 502892848
Encompass_Preferred
Cayman’s SARS-CoV-2 Neutralizing Antibody Detection ELISA Kit is a competitive assay that can be used for qualitative and/or semi-quantitative measurement of neutralizing antibodies in human plasma and serum. When using the neutralizing antibody standard provided in the kit, the assay has a range of 7.81-1,000 ng/ml with a midpoint of approximately 120 ng/ml (50% B/B0) and a sensitivity (80% B/B0) of approximately 46 ng/ml.
Catalog No. 50-289-2848 Supplier Cayman Chemical Supplier No. 50207096 WELL
Cayman’s SARS-CoV-2 Neutralizing Antibody Detection ELISA Kit is a competitive assay that can be used for qualitative and/or semi-quantitative measurement of neutralizing antibodies in human plasma and serum. When using the neutralizing antibody standard provided in the kit, the assay has a range of 7.81-1,000 ng/ml with a midpoint of approximately 120 ng/ml (50% B/B0) and a sensitivity (80% B/B0) of approximately 46 ng/ml.