Learn More
Sino Biological SARS-CoV-2 Omicron (B.1.1.529) Spike RBD Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 40592T62OV100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant SARS-CoV-2 B.1.1.529 (Omicron) Spike RBD Protein, Cat#40592-V08H121 (YP_009724390.1, Arg319-Phe541, with mutation G339D, S371L, S373P, S375F, K417N, N440K, G446S, S477N, T478K, E484A, Q493R, G496S, Q498R, N501Y, Y505H). The specific IgG was purified by SARS-CoV-2 B.1.1.529 (Omicron) Spike RBD affinity chromatography.
Catalog No. 50-228-2072