Learn More
Cayman Chemical SARSCoV2 PapaInlIke Protea
SDP
Catalog No. 502949396
Encompass_Preferred
Source: Active recombinant SARS-CoV-2 N-terminal His-tagged papain-like protease expressed in E. coli • Amino acids: 1,564-1,880 • MW: 36.8 kDa
Catalog No. 50-294-9396 Supplier Cayman Chemical Supplier No. 31817100 UG
Source: Active recombinant SARS-CoV-2 N-terminal His-tagged papain-like protease expressed in E. coli • Amino acids: 1,564-1,880 • MW: 36.8 kDa