Learn More
Epigentek Inc SARS-CoV-2 S Recombinant Antibody (100ul)
SDP
Supplier:  Epigentek Inc A73672100
Encompass_Preferred
HRP Conjugated Recombinant Antibody Raised in Mouse. Immunogen: Recombinant Human Novel Coronavirus Spike glycoprotein (S) (16-685aa). Tested Applications: ELISA. Reactivity: SARS-CoV-2. Alternative Names: S, S1, S1-RBD, Spike glycoprotein.
Catalog No. 50-201-9327