Learn More
Epigentek Inc SARS-CoV-2 S Recombinant Antibody, Biotin Conjulated (100ul)
SDP
Supplier:  Epigentek Inc A73675100
Encompass_Preferred
Unconjugated Recombinant Antibody Raised in Mouse. Immunogen: Recombinant Human Novel Coronavirus Nucleoprotein (N) (1-419aa). Tested Applications: ELISA, WB, GICA. Reactivity: SARS-CoV-2. Alternative Names: Nucleocapsid protein, NC, protein N, N.
Catalog No. 50-201-9333