Learn More
Epigentek Inc SARS-CoV-2 Spike S1 RBD Protein, Human Fc-Fusion, Avi-Tag (1 mg)
SDP
Supplier:  Epigentek Inc E800252
Encompass_Preferred
SARS-CoV-2 Spike protein S1 subunit, receptor-binding domain (RBD), also known as SARS-CoV-2 spike RBD, novel coronavirus spike RBD and nCoV spike RBD, GenBank Accession No. MN_908947.1, a.a. 319-541, fused at the C-terminus of the Fc portion of human IgG1, with a C-terminal Avi-tag™, expressed in a HEK293 cell expression system. MW=54 kDa. This protein runs at a higher MW by SDS-PAGE due to glycosylation.
Catalog No. 50-201-9385