Learn More
Epigentek Inc SARS-CoV-2 Spike S1 RBD Protein, Mouse Fc-fusion (20 µg)
SDP
Supplier:  Epigentek Inc E800261
Encompass_Preferred
Severe acute respiratory Coronavirus 2 Spike Glycoprotein S1 Receptor Binding Domain (SARS-CoV-2 Spike S1 RBD), also known as novel coronavirus spike S1 RBD and nCoV spike S1 RBD, GenBank Accession No. QHD43416.1, a.a. 319-541, with a C-terminal mouse Fc-tag (mFc), expressed in a HEK293 cell expression system. MW=50 kDa. This protein runs at a higher MW by SDS-PAGE due to glycosylation.
Catalog No. 50-201-9386