Learn More
Epigentek Inc SARS-CoV-2 Spike S1 RBD (V367F) Protein, Avi-His-tag (1 mg)
SDP
Catalog No. 502019381
Encompass_Preferred
SARS-CoV-2 Spike S1 receptor-binding domain (RBD), also known as SARS-CoV-2 Spike 1 RBD, novel coronavirus Spike 1 RBD, and nCoV Spike 1 RBD, GenBank Accession No. QHD43416.1, a.a. 319-541 with a V367F mutation and a with C-terminal Avi-His-tag, expressed in a HEK293 cell expression system. MW= 28 kDa. This protein runs at a higher M.W. by SDS-PAGE due to glycosylation.
Catalog No. 50-201-9381 Supplier Epigentek Inc Supplier No. E800232
SARS-CoV-2 Spike S1 receptor-binding domain (RBD), also known as SARS-CoV-2 Spike 1 RBD, novel coronavirus Spike 1 RBD, and nCoV Spike 1 RBD, GenBank Accession No. QHD43416.1, a.a. 319-541 with a V367F mutation and a with C-terminal Avi-His-tag, expressed in a HEK293 cell expression system. MW= 28 kDa. This protein runs at a higher M.W. by SDS-PAGE due to glycosylation.