Learn More
Cayman Chemical SARSCoV2 SpIk Glycoprotin 50ug
SDP
Catalog No. 502991220
Encompass_Preferred
Source: Recombinant protein from the receptor-binding domain (RBD) of SARS-CoV-2 (2019-nCoV) Spike S1 of Wuhan pneumonia virus (MN908947.3) with a C-terminal human IgG1 Fc tag expressed in HEK293 cells • Amino acids: 319-541 • MW: 52.3 kDa
Catalog No. 50-299-1220 Supplier Cayman Chemical Supplier No. 3042950 UG
Source: Recombinant protein from the receptor-binding domain (RBD) of SARS-CoV-2 (2019-nCoV) Spike S1 of Wuhan pneumonia virus (MN908947.3) with a C-terminal human IgG1 Fc tag expressed in HEK293 cells • Amino acids: 319-541 • MW: 52.3 kDa