Learn More
Cayman Chemical SARSCoV2 SpIk Glycoprotin 50uL
SDP
Supplier:  Cayman Chemical 3250350 UL
Encompass_Preferred
A σ receptor ligand with anticancer and antiviral activites binds to σ1 and σ2 receptors (Kis 13 and 0.28 nM respectively in MCF-7 cell membranes) selective for σ1 and σ2 receptors over hERG channels (Ki 1000 nM) inhibits growth of MCF-7 and multidrug-resistant MCF-7/adr cells (IC50s 25 and 15 nM respectively) decreases basal P-gp levels in the same cells reduces SARS-CoV-2 infectivity in Vero E6 cells (IC90 278 nM)
Catalog No. 50-294-8345