Learn More
Sino Biological SARS-COV-2 JN.1 (Omicron) Spike RBD Protein (aa319-529, His Tag)(HPLC-verified)
Supplier:  Sino Biological 40592V08H155100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (JN.1) Spike RBD (YP_009724390.1, with mutation I332V, G339H, K356T, S371F, S373P, S375F, T376A, R403K, D405N, R408S, K417N, N440K, V445H, G446S, N450D, L452W, L455S, N460K, S477N, T478K, N481K, V483del, E484K, F486P, Q498R, N501Y, Y505H) (Arg319-Lys529) was expressed with a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant JN.1).
Catalog No. 50-299-3434