Learn More
Sino Biological SARS-COV-2 KP.2 (Omicron) Spike RBD Protein (His Tag)  (HPLC-verified)
Catalog No. 502993360
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (KP.2) Spike RBD (YP_009724390.1, with mutation I332V, G339H, R346T, K356T, S371F, S373P, S375F, T376A, R403K, D405N, R408S, K417N, N440K, V445H, G446S, N450D, L452W, L455S, F456L, N460K, S477N, T478K, N481K, V483del, E484K, F486P, Q498R, N501Y, Y505H) (Arg319-lys529) was expressed with a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant KP.2).
Catalog No. 50-299-3360 Supplier Sino Biological Supplier No. 40592V08H156100
A DNA sequence encoding the SARS-CoV-2 (KP.2) Spike RBD (YP_009724390.1, with mutation I332V, G339H, R346T, K356T, S371F, S373P, S375F, T376A, R403K, D405N, R408S, K417N, N440K, V445H, G446S, N450D, L452W, L455S, F456L, N460K, S477N, T478K, N481K, V483del, E484K, F486P, Q498R, N501Y, Y505H) (Arg319-lys529) was expressed with a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant KP.2).