Learn More
Sino Biological SARS-COV-2 JN.1 (Omicron) Spike RBD Protein (aa319-529, His & AVI Tag), Biotinylated(HPLC-verified)
Catalog No. 502993467
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike RBD (YP_009724390.1, with mutation I332V, G339H, K356T, S371F, S373P, S375F, T376A, R403K, D405N, R408S, K417N, N440K, V445H, G446S, N450D, L452W, L455S, N460K, S477N, T478K, N481K, V483del, E484K, F486P, Q498R, N501Y, Y505H) (Arg319-Lys529) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed. The mutations were identified in the SARS-CoV-2 variant (known as variant JN.1).
Catalog No. 50-299-3467 Supplier Sino Biological Supplier No. 40592V49H16B20
A DNA sequence encoding the SARS-CoV-2 Spike RBD (YP_009724390.1, with mutation I332V, G339H, K356T, S371F, S373P, S375F, T376A, R403K, D405N, R408S, K417N, N440K, V445H, G446S, N450D, L452W, L455S, N460K, S477N, T478K, N481K, V483del, E484K, F486P, Q498R, N501Y, Y505H) (Arg319-Lys529) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed. The mutations were identified in the SARS-CoV-2 variant (known as variant JN.1).