Learn More
Sino Biological SARS-CoV-2 Spike Antibody (Biotin), Rabbit PAb, Antigen Affinity Purified 100ug
Supplier:  Sino Biological 40589T62B100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant SARS-CoV-2 / 2019-nCoV Spike Protein (S1+S2 ECD)(Catalog#40589-V08B1; YP_009724390.1; Val16-Pro1213). The specific IgG was purified by SARS-CoV-2 / 2019-nCoV Spike affinity chromatography and then biotinylated.
Catalog No. 50-224-6471