Learn More
Sino Biological SARS-CoV-2 Spike Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 40592R190100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant SARS-CoV-2 Spike ( Catalog#40592-V05H; YP_009724390.1; Arg319-Phe541).
Catalog No. 50-224-7378