Learn More
Sino Biological SARS-CoV-2 Spike Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 40591T30100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant SARS-CoV-2 Spike ( Catalog#40591-V08H; YP_009724390.1; Val16-Arg685). Spike specific IgG was purified by SARS-CoV-2 Spike affinity chromatography.
Catalog No. 50-224-6486