Learn More
Cayman Chemical SARSCoV2 SpIke GlycoproteIn
SDP
Supplier:  Cayman Chemical 31803100 UG
Encompass_Preferred
An activator of Nrf2 signaling increases the expression of Nrf2 target genes and activates HO-1 in CV-1 cells at 100 nM binds to PPARY and PPARa (Kis 344 and 232 nM respectively) and induces PPARY transactivation in SW480 cells inhibits NOS production in isolated mouse macrophages (IC50 0.014 nM) inhibits proliferation of U937 and MCF-7 cells at 300 nM reduces fatty acid synthesis in LiSa-2 human liposarcoma cells at 100 nM reduces tumor growth in a B16 murine melanoma model at 50 Ug/animal twice per day
Catalog No. 50-294-7049