Learn More
Cayman Chemical SARSCoV2 SpIke GlycoproteIn
SDP
Supplier:  Cayman Chemical 31998100 UL
Encompass_Preferred
A cGAS inhibitor (IC50 0.11 UM) inhibits dsDNA-induced IFN-B1-dependent gene expression in a reporter assay reduces basal Ifnb1 mRNA expression in BMDMs generated from the Trex1-/- mouse model of Aicardi-Goutières syndrome
Catalog No. 50-294-8420