Learn More
Cayman Chemical SARSCoV2 SpIke GlycoproteIn
SDP
Supplier:  Cayman Chemical 31814100 UG
Encompass_Preferred
Source: Active recombinant C-terminal mouse IgG1 Fc-tagged SARS-CoV-2 surface glycoprotein S1 subunit expressed in HEK293 cells • Amino acids: 16-685 • MW: 101.4 kDa
Catalog No. 50-294-9090