Learn More
Cayman Chemical SARSCoV2 SpIke GlycoproteIn
Supplier: Cayman Chemical 31997100 UL
A cGAS inhibitor (IC50 0.11 UM) inhibits dsDNA-induced IFN-B1-dependent gene expression in a reporter assay reduces basal Ifnb1 mRNA expression in BMDMs generated from the Trex1-/- mouse model of Aicardi-Goutières syndrome