Learn More
Cayman Chemical SARSCoV2 SpIke GlycoproteIn
SDP
Supplier:  Cayman Chemical 32571100 UG
Encompass_Preferred
Source: Active recombinant SARS-CoV-2 C-terminal His-tagged spike glycoprotein receptor binding domain expressed in HEK293 cells • Amino acids: 21-243, 319-541 of P0DTC2 • MW: 28.7 kDa
Catalog No. 50-294-9366