Learn More
Cayman Chemical SARSCoV2 SpIke GlycoproteIn
SDP
Supplier:  Cayman Chemical 32501100 UG
Encompass_Preferred
A BD2 bromodomain inhibitor inhibits BD2 in BRD2 BRD3 BRD4 and BRDT (IC50s 264 98 49 and 214 nM respectively) and is selective for BD2 over BD1 (IC50s 10.96 36.31 70.55 and 50.11 UM for BRD2 BRD3 BRD4 and BDT respectively) in a TR-FRET assay inhibits IFN-Y-induced protein expression of MHC class I in K562 cells at 1 UM reduces the production of anti-KLH IgM antibodies in mice immunized with KLH at 40 mg/kg
Catalog No. 50-294-7382