Learn More
Cayman Chemical SARSCoV2 SpIke GlycoproteIn
SDP
Supplier:  Cayman Chemical 31815100 UG
Encompass_Preferred
Source: Recombinant C-terminal mouse IgG1 Fc-tagged SARS-CoV-2 surface glycoprotein S2 subunit expressed in insect cells • Amino acids: 686-1,213 • MW: 84.3 kDa
Catalog No. 50-294-7783