Learn More
Cayman Chemical SARSCoV2 SpIke GlycoproteIn
SDP
Supplier:  Cayman Chemical 33867100 UG
Encompass_Preferred
Source: Active recombinant C-terminal rabbit IgG1 Fc-tagged SARS-CoV-2 surface glycoprotein receptor binding domain N501Y variant expressed in HEK293 cells • Amino acids: 21-243, 319-541 of P0DTC2 • MW: 50.37 kDa
Catalog No. 50-294-7478