Learn More
Cayman Chemical SARSCoV2 SpIke GlycoproteIn
Supplier: Cayman Chemical 33868100 UG
Source: Active recombinant C-terminal rabbit IgG1 Fc-tagged SARS-CoV-2 Spike Glycoprotein Receptor Binding Domain K417N, E484K, N501Y variant expressed in HEK293 cells • Amino acids: 21-243, 319-541 of P0DTC2 • MW: 50.35 kDa