Learn More
Cayman Chemical SARSCoV2 SpIke GlycoproteIn
SDP
Supplier:  Cayman Chemical 31994100 UL
Encompass_Preferred
An ENPP1 inhibitor inhibits the hydrolysis of 23-cGAMP and AMP-pNP (Kis ≤100 and 100-1000 nM respectively in enzyme assays) enhances transcription of the gene encoding IFN-B in 23-cGAMP-stimulated HFF-1 cells
Catalog No. 50-294-7910