Learn More
Cayman Chemical SARSCoV2 SpIke GlycoproteIn
SDP
Supplier:  Cayman Chemical 34837100 UG
Encompass_Preferred
Source: Active recombinant C-terminal rabbit IgG1 Fc-tagged SARS-CoV-2 Spike glycoprotein receptor binding domain L452R, T478K variant expressed in HEK293 cells • Amino acids: 21-243, 319-541 • MW: 50.4 kDa
Catalog No. 50-291-1291