Learn More
Sino Biological SARS-CoV-2 Spike S2 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 40590MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, Recombinant SARS-CoV-2 Spike S2 Protein ( Catalog# 40590-V08B; YP_009724390.1; Ser686-Pro1213). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-222-1608