Learn More
Sino Biological SARS-CoV-2 Spike RBD Antibody (HRP), Rabbit PAb, Antigen Affinity Purified 100ug
Supplier:  Sino Biological 40592T62H100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant SARS-CoV-2 / 2019-nCoV Spike/RBD Protein (YP_009724390.1; Arg319-Phe541). The specific IgG was purified by SARS-CoV-2 / 2019-nCoV Spike/RBD affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-224-7383