Learn More
Sino Biological SARS-CoV-2 Spike RBD (E484D) Protein (His Tag)
Supplier:  Sino Biological 40592V08H104100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike RBD (YP_009724390.1, with mutation E484D) (Arg319-Phe541) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-4311